2-Hydroxy-4-(trifluormethyl)pyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(Trifluormethyl)pyridin-2-ol

M-Clarity™ Qualitätsstufe = MQ100

648876-250MG
SIAL648876-250MGEA 76.6 EUR
SIAL648876-250MG
2-Hydroxy-4-(trifluormethyl)pyridin, Sigma-Aldrich®
2-Hydroxy-4-(trifluoromethyl)pyridine
Formel: C₆H₄F₃NO
Molecular Weight: 163,1 g/mol
MDL: MFCD00153195
CAS-Nummer: 50650-59-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR