2-Hydroxy-6-methylisonicotinsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Hydroxy-6-methylpyridine-4-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

649449-10G
SIAL649449-10GEA 271 EUR
SIAL649449-10G
2-Hydroxy-6-methylisonicotinsäure, Sigma-Aldrich®
2-Hydroxy-6-methylisonicotinsäure
Formel: C₇H₇NO₃
Molecular Weight: 153,15 g/mol
MDL: MFCD02682019
CAS-Nummer: 86454-13-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR