4-(3-Thienyl)anilin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

650080-1G
SIAL650080-1GEA 90.3 EUR
SIAL650080-1G
4-(3-Thienyl)anilin, Sigma-Aldrich®
4-(3-Thienyl)anilin
Formel: C₁₀H₉NS
MDL: MFCD06740168
CAS-Nummer: 834884-74-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR