Methyl 4-(4-formylphenyl)benzoate, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4'-Formylbiphenyl-4-carbonsäuremethylester

M-Clarity™ Qualitätsstufe = MQ100

650099-1G
SIAL650099-1GEA 90.8 EUR
SIAL650099-1G
Methyl 4-(4-formylphenyl)benzoate, Sigma-Aldrich®
Methyl 4-(4-formylphenyl)benzoate
Formel: C₁₅H₁₂O₃
Molecular Weight: 240.25 g/mol
MDL: MFCD04039123
CAS-Nummer: 70916-89-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR