1-(4-tert-Butylbenzyl)piperazin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

650129-1G
SIAL650129-1GEA 100 EUR
SIAL650129-1G
1-(4-tert-Butylbenzyl)piperazin, Sigma-Aldrich®
1-(4-tert-Butylbenzyl)piperazin
Formel: C₁₅H₂₄N₂
Molecular Weight: 232,37 g/mol
MDL: MFCD00082594
CAS-Nummer: 956-61-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR