Benzofurazan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,1,3-Benzoxadiazole, Benzo[c][1,2,5]oxadiazole

650137-1G
SIAL650137-1GEA 30.5 EUR
SIAL650137-1G
Benzofurazan, Sigma-Aldrich®
Benzofurazan
Formel: C₆H₄N₂O
Molecular Weight: 120,11 g/mol
MDL: MFCD00068064
CAS-Nummer: 273-09-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR