1-(2-Fluorbenzyl)piperazin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

650242-5G
SIAL650242-5GEA 94.5 EUR
SIAL650242-5G
1-(2-Fluorbenzyl)piperazin, Sigma-Aldrich®
1-(2-Fluorbenzyl)piperazin
Formel: C₁₁H₁₅FN₂
Molecular Weight: 194,25 g/mol
MDL: MFCD02256027
CAS-Nummer: 89292-78-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR