1-Boc-3-piperidon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-BOC-3-piperidone, 1-Boc-3-Piperidinone

M-Clarity™ Qualitätsstufe = MQ100

650811-25G 650811-5G
SIAL650811-25GEA 54.2 EUR
SIAL650811-25G SIAL650811-5G
1-Boc-3-piperidon, Sigma-Aldrich®
1-Boc-3-piperidon
Formel: C₁₀H₁₇NO₃
MDL: MFCD01631193
CAS-Nummer: 98977-36-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR