4-Amino-3-nitrophenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Amino-3-nitrophenylboronicacid

M-Clarity™ Qualitätsstufe = MQ100

651621-1G
SIAL651621-1GEA 311 EUR
SIAL651621-1G
4-Amino-3-nitrophenylboronsäure, Sigma-Aldrich®
4-Amino-3-nitrophenylboronsäure
Formel: C₆H₇BN₂O₄
Molecular Weight: 181,9418 g/mol
MDL: MFCD07437851
CAS-Nummer: 89466-07-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR