4-Fluorphenethylbromid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Fluoro-4-(2-bromoethyl)benzene, 1-(2-bromoethyl)-4-fluorobenzene

652008-5G
SIAL652008-5GEA 79.9 EUR
SIAL652008-5G
4-Fluorphenethylbromid, Sigma-Aldrich®
4-Fluorphenethylbromid
Formel: C₈H₈BrF
Molecular Weight: 203,05 g/mol
MDL: MFCD03844794
CAS-Nummer: 332-42-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR