Hydrotalcite (synthetisch), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dialuminum hexamagnesium carbonate hexadecahydroxide hydrate, Magnesium aluminum hydroxide carbonate hydrate, Magnesium aluminum hydroxycarbonate hydrate

M-Clarity™ Qualitätsstufe = MQ100

652288-1KG
SIAL652288-1KGEA 121 EUR
SIAL652288-1KG
Hydrotalcite (synthetisch), Sigma-Aldrich®
Hydrotalcite

Not easily dispersed in water; has excellent dispersibility in polymer solutions.

Formel: CH₁₆Al₂Mg₆O₁₉.4H₂O
Molecular Weight: 603,973 g/mol (anhy)
Dichte: 2,0 g/mL (25 °C) (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD01746913
CAS-Nummer: 11097-59-9

Bestellen

Spezifikation Testergebnisse

Appearance (Color) White
Appearance (Form) Powder
X-Ray Diffraction Conforms to Structure
Ratio MgO/Al2O3 4.0 - 5.0
Loss on Drying < 0.35 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR