Methyl-5-brom-2-iodbenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 5-bromo-2-iodobenzoate

M-Clarity™ Qualitätsstufe = MQ100

652601-10G
SIAL652601-10GEA 151 EUR
SIAL652601-10G
Methyl-5-brom-2-iodbenzoat, Sigma-Aldrich®
Methyl-5-brom-2-iodbenzoat
Formel: C₈H₆BrIO₂
Molecular Weight: 340,9400 g/mol
MDL: MFCD00144771
CAS-Nummer: 181765-86-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR