4-Fluor-m-anisaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Fluor-3-anisaldehyd, 4-Fluor-3-methoxybenzaldehyd

M-Clarity™ Qualitätsstufe = MQ200

652687-5G
SIAL652687-5GEA 40.5 EUR
SIAL652687-5G
4-Fluor-m-anisaldehyd, Sigma-Aldrich®
4-Fluor-m-anisaldehyd
Formel: C₈H₇FO₂
Molecular Weight: 154.14 g/mol
MDL: MFCD00143320
CAS-Nummer: 128495-46-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR