4-Fluor-o-phenylendiamin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-Fluorbenzol-1,2-diamin, 1,2-Diamino-4-fluorbenzol, 3,4-Diamino-1-fluorbenzol, 4-Fluor-1,2-phenylendiamin
653586-5G
SIAL653586-5GEA
120
EUR
SIAL653586-5G
4-Fluor-o-phenylendiamin, Sigma-Aldrich®
4-Fluor-o-phenylendiamin
Formel:
C₆H₇FN₂ Molecular Weight: 126,13 g/mol Siedepunkt: 250 °C (1013 hPa) Schmelzpunkt: 92 °C |
MDL:
MFCD00042228 CAS-Nummer: 367-31-7 EINECS: 206-691-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...