1-Methyl-1H-pyrazol-4-carbaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Methyl-1H-pyrazole-4-carbaldehyde, 1-Methyl-1H-pyrazole-4-carboxaldehyde

M-Clarity™ Qualitätsstufe = MQ100

654043-1G
SIAL654043-1GEA 33 EUR
SIAL654043-1G
1-Methyl-1H-pyrazol-4-carbaldehyd, Sigma-Aldrich®
1-Methyl-1H-pyrazol-4-carbaldehyd
Formel: C₅H₆N₂O
Molecular Weight: 110,12 g/mol
MDL: MFCD00460465
CAS-Nummer: 25016-11-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR