2,6-Dichlor-4-iodpyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

654078-1G
SIAL654078-1GEA 57.2 EUR
SIAL654078-1G
2,6-Dichlor-4-iodpyridin, Sigma-Aldrich®
2,6-Dichloro-4-iodopyridine
Formel: C₅H₂CI₂IN
Molecular Weight: 273,89 g/mol
MDL: MFCD07368400
CAS-Nummer: 98027-84-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR