4-Iod-2-nitroanilin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

654108-1G
SIAL654108-1GEA 0 EUR
SIAL654108-1G
4-Iod-2-nitroanilin, Sigma-Aldrich®
4-Iod-2-nitroanilin
Formel: C₆H₅IN₂O₂
MDL: MFCD06808526
CAS-Nummer: 20691-72-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR