4-(2-Thienyl)anilin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(Thiophen-2-yl)aniline, 4-Thiophen-2-ylphenylamine

M-Clarity™ Qualitätsstufe = MQ100

654558-1G
SIAL654558-1GEA 104 EUR
SIAL654558-1G
4-(2-Thienyl)anilin, Sigma-Aldrich®
4-(2-Thienyl)anilin
Formel: C₁₀H₉NS
MDL: MFCD01224064
CAS-Nummer: 70010-48-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR