ω,3-Dibromacetophenon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2,3'-Dibromacetophenon, 2-Brom-1-(3-bromphenyl)-1-ethanon, 3-Bromphenacylbromid, α,3-Dibromacetophenon
M-Clarity™ Qualitätsstufe = MQ100
654787-5G
SIAL654787-5GEA
36.3
EUR
SIAL654787-5G
ω,3-Dibromacetophenon, Sigma-Aldrich®
ω,3-Dibromacetophenon
Formel:
C₈H₆Br₂O Molecular Weight: 277,94 g/mol Schmelzpunkt: 51 °C |
MDL:
MFCD00082698 CAS-Nummer: 18523-22-3 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...