1-(2,5-Difluorphenyl)-2-thioharnstoff, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2,5-Difluorphenylthioharnstoff, N-(2,5-Difluorphenyl)thioharnstoff
M-Clarity™ Qualitätsstufe = MQ100
654868-1G
SIAL654868-1GEA
32.4
EUR
SIAL654868-1G
1-(2,5-Difluorphenyl)-2-thioharnstoff, Sigma-Aldrich®
1-(2,5-Difluorphenyl)-2-thioharnstoff
Formel:
(C₆H₃F₂)NC(S)NH₂ Molecular Weight: 188,2 g/mol Siedepunkt: 272 °C (1013 hPa) Schmelzpunkt: 137…138 °C |
MDL:
MFCD00041152 CAS-Nummer: 207981-44-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...