Tri-tert-butylphosphin 1 mol/l in Toluol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

655325-10ML 655325-50ML
SIAL655325-10MLEA 132 EUR
SIAL655325-10ML SIAL655325-50ML
Tri-tert-butylphosphin 1 mol/l in Toluol, Sigma-Aldrich®
Tri-tert-butylphosphin
Formel: C₁₂H₂₇P
MDL: MFCD00015006
CAS-Nummer: 13716-12-6
EINECS: 237-266-4
UN: 2845
ADR: 4.2,I

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR