3,4'-Methylendianilin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,4'-Diaminodiphenylmethane, 3,4'-Methylenedianiline

M-Clarity™ Qualitätsstufe = MQ100

655449-1G
SIAL655449-1GEA 64.3 EUR
SIAL655449-1G
3,4'-Methylendianilin, Sigma-Aldrich®
3,4'-Methylendianilin
Formel: C₁₃H₁₄N₂
MDL: MFCD00014778
CAS-Nummer: 19430-83-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR