2-Acrylamido-2-methylpropansulfonsäure Natriumsalz 50% aqueous solution, Sigma-Aldrich®

Lieferant: Merck


MQ200

655821-1L 655821-250ML
SIAL655821-1LEA 203 EUR
SIAL655821-1L SIAL655821-250ML
2-Acrylamido-2-methylpropansulfonsäure Natriumsalz 50% aqueous solution, Sigma-Aldrich®
2-Acrylamido-2-methylpropansulfonsäure Natriumsalz
Formel: C₇H₁₂NNaO₄S
MDL: MFCD00156481
CAS-Nummer: 5165-97-9
EINECS: 225-948-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR