Methyl-4-amino-3-brombenzoat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Methyl 4-amino-3-bromobenzoate
M-Clarity™ Qualitätsstufe = MQ200
655910-5G
SIAL655910-5GEA
84.5
EUR
SIAL655910-5G
Methyl-4-amino-3-brombenzoat, Sigma-Aldrich®
Methyl-4-amino-3-brombenzoat
Formel:
C₈H₈BrNO₂ Molecular Weight: 230,06 g/mol |
MDL:
MFCD01861385 CAS-Nummer: 106896-49-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...