4-Brom-3,5-dichlorbenzotrifluorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

655996-1G
SIAL655996-1GEA 106 EUR
SIAL655996-1G
4-Brom-3,5-dichlorbenzotrifluorid, Sigma-Aldrich®
4-Brom-3,5-dichlorbenzotrifluorid
Formel: C₇H₂BrCl₂F₃
Molecular Weight: 293,9 g/mol
MDL: MFCD00221020
CAS-Nummer: 118754-53-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR