Methyl-2,6-dichlornicotinat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 2,6-dichloronicotinate

M-Clarity™ Qualitätsstufe = MQ100

656437-1G
SIAL656437-1GEA 82.2 EUR
SIAL656437-1G
Methyl-2,6-dichlornicotinat, Sigma-Aldrich®
Methyl-2,6-dichlornicotinat
Formel: C₇H₅Cl₂NO₂
Molecular Weight: 206,03 g/mol
MDL: MFCD07369794
CAS-Nummer: 65515-28-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR