1,3-Diisopropylimidazoliumchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,3-Di-i-propylimidazolium chloride, 1,3-Diisopropyl-1H-imidazol-3-ium chloride

M-Clarity™ Qualitätsstufe = MQ200

656577-1G 656577-5G
SIAL656577-1GEA 62.6 EUR
SIAL656577-1G SIAL656577-5G
1,3-Diisopropylimidazoliumchlorid, Sigma-Aldrich®
1,3-Diisopropylimidazoliumchlorid
Formel: C₉H₁₇ClN₂
Molecular Weight: 188.70 g/mol
MDL: MFCD03840577
CAS-Nummer: 139143-09-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR