2-Amino-4,6-dichlorpyrimidin-5-carbaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Amino-4,6-dichloropyrimidine-5-carboxaldehyde

M-Clarity™ Qualitätsstufe = MQ100

656720-1G
SIAL656720-1GEA 68.9 EUR
SIAL656720-1G
2-Amino-4,6-dichlorpyrimidin-5-carbaldehyd, Sigma-Aldrich®
2-Amino-4,6-dichlorpyrimidin-5-carbaldehyd
Formel: C₅H₃Cl₂N₃O
Molecular Weight: 192,00 g/mol
MDL: MFCD03001242
CAS-Nummer: 5604-46-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR