4-Cyan-2-iodanilin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Amino-3-iodbenzoesäurenitril, 4-Amino-3-iodbenzonitril

657123-5G
SIAL657123-5GEA 80.7 EUR
SIAL657123-5G
4-Cyan-2-iodanilin, Sigma-Aldrich®
4-Cyan-2-iodanilin
Formel: C₇H₅IN₂
Molecular Weight: 244.03 g/mol
MDL: MFCD04039964
CAS-Nummer: 33348-34-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR