3-(3,4-Methylendioxy)phenylpropionsäure ≥97%, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
3,4-(Methylenedioxy)hydrocinnamic acid, 3-(1,3-Benzodioxyl-5-yl)propionic acid
M-Clarity™ Qualitätsstufe = MQ100
657565-1G
SIAL657565-1GEA
31.5
EUR
SIAL657565-1G
3-(3,4-Methylendioxy)phenylpropionsäure ≥97%, Sigma-Aldrich®
3-(3,4-Methylendioxy)phenylpropionsäure
Formel:
C₁₀H₁₀O₄ Molecular Weight: 194,19 g/mol Schmelzpunkt: 83…87 °C |
MDL:
MFCD00016909 CAS-Nummer: 2815-95-4 EINECS: 220-565-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...