1-(4-(Trifluormethyl)benzyl)piperazin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

657743-1G
SIAL657743-1GEA 55.2 EUR
SIAL657743-1G
1-(4-(Trifluormethyl)benzyl)piperazin, Sigma-Aldrich®
1-(4-(Trifluormethyl)benzyl)piperazin
Formel: C₁₂H₁₅F₃N₂
MDL: MFCD03407490
CAS-Nummer: 107890-32-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR