Methyl-N-methylanthranilat, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: N-Methylanthranilsäuremethylester, Dimethylanthranilat, 2-(Methylamino)benzoesäuremethylester, N-Methylanthranilic acid methyl ester

65820-100ML
SIAL65820-100MLEA 0 EUR
SIAL65820-100ML
Methyl-N-methylanthranilat, Sigma-Aldrich®
Methyl-N-methylanthranilat
Formel: C₉H₁₁NO₂
Molecular Weight: 165,19 g/mol
Dichte: 1,12 g/cm³ (15 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00017183
CAS-Nummer: 85-91-6
EINECS: 201-642-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR