2-Methoxy-3-(trifluormethyl)pyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

658359-5G
SIAL658359-5GEA 75.4 EUR
SIAL658359-5G
2-Methoxy-3-(trifluormethyl)pyridin, Sigma-Aldrich®
2-Methoxy-3-(trifluoromethyl)pyridine
Formel: C₇H₆F₃NO
Molecular Weight: 177,13 g/mol
MDL: MFCD00153202
CAS-Nummer: 121643-44-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR