3,5-Dibrom-p-toluidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,5-Dibromo-4-methylaniline

M-Clarity™ Qualitätsstufe = MQ100

658367-5G
SIAL658367-5GEA 117 EUR
SIAL658367-5G
3,5-Dibrom-p-toluidin, Sigma-Aldrich®
3,5-Dibrom-p-toluidin
Formel: C₇H₇Br₂N
Molecular Weight: 264,96 g/mol
MDL: MFCD00151806
CAS-Nummer: 13194-73-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR