5-Brom-2-hydroxy-3-nitropyridin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
658448-25G
SIAL658448-25GEA
123
EUR
SIAL658448-25G
5-Brom-2-hydroxy-3-nitropyridin, Sigma-Aldrich®
5-Brom-2-hydroxy-3-nitropyridin
Formel:
C₅H₃BrN₂O₃ |
MDL:
MFCD00023473 CAS-Nummer: 15862-34-7 EINECS: 239-989-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...