4-Methyl-1,10-phenanthrolin, Sigma-Aldrich®

Lieferant: Merck

658529-1G
SIAL658529-1GEA 105 EUR
SIAL658529-1G
4-Methyl-1,10-phenanthrolin, Sigma-Aldrich®
4-Methyl-1,10-phenanthroline
Formel: C₁₃H₁₀N₂
MDL: MFCD00004978
CAS-Nummer: 31301-28-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR