2-(4-Bromphenyl)-6-methylimidazo[1,2-a]pyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

658960-1G
SIAL658960-1GEA 76.4 EUR
SIAL658960-1G
2-(4-Bromphenyl)-6-methylimidazo[1,2-a]pyridin, Sigma-Aldrich®
2-(4-Bromphenyl)-6-methylimidazo[1,2-a]pyridin
Formel: C₁₄H₁₁BrN₂
Molecular Weight: 287,16 g/mol
MDL: MFCD00444784
CAS-Nummer: 1023-01-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR