2-(4-Bromphenyl)imidazo[1,2-a]pyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

658979-1G
SIAL658979-1GEA 30.9 EUR
SIAL658979-1G
2-(4-Bromphenyl)imidazo[1,2-a]pyridin, Sigma-Aldrich®
2-(4-Bromphenyl)imidazo[1,2-a]pyridin
Formel: C₁₃H₉BrN₂
MDL: MFCD00218250
CAS-Nummer: 34658-66-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR