Carbon nanotube mehrwandig ≥90% (Carbon-Basis), Pulver, Länge 5 - 9 µm, Ø 110 - 170 nm, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: CNT

M-Clarity™ Qualitätsstufe = MQ100

659258-10G
SIAL659258-10GEA 477 EUR
SIAL659258-10G
Carbon nanotube mehrwandig ≥90% (Carbon-Basis), Pulver, Länge 5 - 9 µm, Ø 110 - 170 nm, Sigma-Aldrich®
Carbon nanotube

Multiwalled carbon nanotubes were used to design a 3D, free standing porous carbon structure.


  • Design for energy efficiency


Toxic and immunological effects of carbon nanotubes on human colorectal cancer cell line (CaCO⁻²), the human breast adenocarcinoma cell line (MCF-7, calu-3 cells from human lung adenocarcinoma and serous cells of proximal bronchial airways was investigated. The self-supporting structure of carbon nanotube could find potential uses in energy storage, supercapacitors, photovoltaic cells, field emission devices, sensors and high performance catalysts. PS/MWNTs nanocomposites have been fabricated and studied for its electrical and optical properties.


Multi walled carbon nanotubes (MWNTs, CNTs) were prepared by chemical vapor deposition (CVD). In chemical vapor deposition (CVD), a volatile precursor undergoes thermal decomposition at elevated temperatures to form a solid deposit on a substrate.


Synonym: MWCNT, MWNT, multiwall carbon nanotube.

Formel: C
Molecular Weight: 12,011 g/mol
Schmelzpunkt: 3652…3697 °C (Lit.)
Dichte: 1,7 g/ml (25 °C)
MDL: MFCD00133992
CAS-Nummer: 308068-56-6

Bestellen

Spezifikation Testergebnisse

Assay 90%
Form Powder
Composition carbon >90%
Diameter 110 – 170 nm
Density (25 °C) 1,7 g/ml
Color Black
Iron (Fe) ≤0,1%
Length 5 – 9 μm
Stabilty (inert atmosphere) 3697 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR