1,3-Bis(1-adamantyl)imidazoliumtetrafluoroborat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1,3-Di(adamantan-1-yl)-1H-imidazol-3-ium tetrafluoroborate
M-Clarity™ Qualitätsstufe = MQ100
660035-1G
SIAL660035-1GEA
167
EUR
SIAL660035-1G
1,3-Bis(1-adamantyl)imidazoliumtetrafluoroborat, Sigma-Aldrich®
1,3-Bis(1-adamantyl)imidazoliumtetrafluoroborat
Formel:
C₂₃H₃₃BF₄N₂ Molecular Weight: 424,33 g/mol |
MDL:
MFCD04973311 CAS-Nummer: 286014-42-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...