4-Chlornicotinsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Chlor-3-pyridincarbonsäure, 4-Chlorpyridin-3-carbonsäure

M-Clarity™ Qualitätsstufe = MQ200

660396-1G
SIAL660396-1GEA 35.9 EUR
SIAL660396-1G
4-Chlornicotinsäure, Sigma-Aldrich®
4-Chlornicotinsäure
Formel: C₆H₄ClNO₂
Molecular Weight: 157,56 g/mol
MDL: MFCD00128860
CAS-Nummer: 10177-29-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR