2,3,6-Trifluorisonicotinsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,3,6-Trifluoropyridine-4-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

660639-500MG
SIAL660639-500MGEA 39.5 EUR
SIAL660639-500MG
2,3,6-Trifluorisonicotinsäure, Sigma-Aldrich®
2,3,6-Trifluorisonicotinsäure
Formel: C₆H₂F₃NO₂
Molecular Weight: 177,08 g/mol
MDL: MFCD03840204
CAS-Nummer: 675602-92-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR