2-Brom-4-(trifluormethyl)pyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

661139-500MG
SIAL661139-500MGEA 56.7 EUR
SIAL661139-500MG
2-Brom-4-(trifluormethyl)pyridin, Sigma-Aldrich®
2-Bromo-4-(trifluoromethyl)pyridine
Formel: C₆H₃BrF₃N
Molecular Weight: 226 g/mol
MDL: MFCD00153085
CAS-Nummer: 175205-81-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR