2-Brom-6-(trifluormethyl)pyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

661147-500MG
SIAL661147-500MGEA 76.6 EUR
SIAL661147-500MG
2-Brom-6-(trifluormethyl)pyridin, Sigma-Aldrich®
2-Bromo-6-(trifluoromethyl)pyridine
Formel: C₆H₃BrF₃N
Molecular Weight: 226 g/mol
MDL: MFCD00153087
CAS-Nummer: 189278-27-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR