3-Amino-2-brompyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Brom-3-pyridylamin, 2-Brompyridin-3-amin

M-Clarity™ Qualitätsstufe = MQ100

661228-1G
SIAL661228-1GEA 35.2 EUR
SIAL661228-1G
3-Amino-2-brompyridin, Sigma-Aldrich®
3-Amino-2-brompyridin
Formel: C₅H₅BrN₂
Molecular Weight: 173,01 g/mol
MDL: MFCD00234064
CAS-Nummer: 39856-58-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR