Vinyltrimethylsilan, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Trimethyl(vinyl)silane
M-Clarity™ Qualitätsstufe = MQ100
661724-25G
SIAL661724-25GEA
0
EUR
SIAL661724-25G
Vinyltrimethylsilan, Sigma-Aldrich®
Vinyltrimethylsilan
Formel:
C₅H₁₂Si Molecular Weight: 100,24 g/mol Siedepunkt: 55 °C (1013 hPa) Dichte: 0,7038 g/cm³ (20 °C) |
MDL:
MFCD00008606 CAS-Nummer: 754-05-2 EINECS: 212-042-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...