2-Pyridinsulfonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Pyridine-2-sulphonic acid

M-Clarity™ Qualitätsstufe = MQ200

661759-10G
SIAL661759-10GEA 302 EUR
SIAL661759-10G
2-Pyridinsulfonsäure, Sigma-Aldrich®
2-Pyridinesulphonic acid
Formel: C₅H₅NO₃S
Molecular Weight: 159,1631 g/mol
MDL: MFCD00234080
CAS-Nummer: 15103-48-7
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR