3-Brom-2-methylpyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Brom-2-picolin

M-Clarity™ Qualitätsstufe = MQ100

662720-1G
SIAL662720-1GEA 35.4 EUR
SIAL662720-1G
3-Brom-2-methylpyridin, Sigma-Aldrich®
3-Brom-2-methylpyridin
Formel: C₆H₆BrN
Molecular Weight: 172.02 g/mol
MDL: MFCD00191224
CAS-Nummer: 38749-79-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR