2-Morpholinoanilin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(4-Morpholinyl)aniline

M-Clarity™ Qualitätsstufe = MQ100

663204-5G
SIAL663204-5GEA 82 EUR
SIAL663204-5G
2-Morpholinoanilin, Sigma-Aldrich®
2-Morpholinoanilin
Formel: C₁₀H₁₄N₂O
Molecular Weight: 178,23 g/mol
MDL: MFCD00047408
CAS-Nummer: 5585-33-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR