2,6-Difluor-4-formylphenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,6-Difluoro-4-formylphenylboronic acid, pinacol ester, 3,5-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

M-Clarity™ Qualitätsstufe = MQ100

663514-5G
SIAL663514-5GEA 419 EUR
SIAL663514-5G
2,6-Difluor-4-formylphenylboronsäurepinakolester, Sigma-Aldrich®
2,6-Difluor-4-formylphenylboronsäurepinakolester
Formel: C₁₃H₁₅BF₂O₃
Molecular Weight: 268.600 g/mol
MDL: MFCD08276848
CAS-Nummer: 870717-92-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR